Please login to your online account to display your discounted pricing

anti-p53, Clone: PAb122, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2328921ML

 View all options for this product


For Research Use Only

Catalog No. NBP2328921M


Description & Specifications


Antigen p53
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Clone PAb122
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P04637
Host Species Mouse
Immunogen SV40-transformed mouse B4 cells
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7157