Please login to your online account to display your discounted pricing

anti-p53, Clone: PAb122, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232892

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232892

  • / Each
 

Description & Specifications

Specifications

Antigen p53
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Clone PAb122
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Hamster
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P04637
Host Species Mouse
Immunogen SV40-transformed mouse B4 cells
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7157