Please login to your online account to display your discounted pricing

anti-p53, Clone: PAb122, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone PAb122
Host Species Mouse
Isotype κ
Immunogen SV40-transformed mouse B4 cells
Antigen p53
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232892

 
novus biologicals canada
NBP232892
Unlabeled 0.5mL Each for $321.00

NBP2328921M

 
novus biologicals canada
NBP2328921ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone PAb122
Host Species Mouse
Isotype κ
Immunogen SV40-transformed mouse B4 cells
Antigen p53
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P04637
Gene ID (Entrez) 7157
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL