Please login to your online account to display your discounted pricing

anti-p27/Kip1, Clone: SX53G8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328891ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328891M

  • / Each
 

Description & Specifications

Specifications

Antigen p27/Kip1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SX53G8
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P46527
Host Species Mouse
Immunogen Purified GST-p27 fusion protein of human origin
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 1027