Please login to your online account to display your discounted pricing

anti-p27/Kip1, Clone: SX53G8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232889

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232889

  • / Each
 

Description & Specifications

Specifications

Immunogen Purified GST-p27 fusion protein of human origin
Conjugate Unlabeled
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Isotype κ
Clone SX53G8
Regulatory Status RUO
Antigen p27/Kip1
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Gene Accession No. P46527
Gene ID (Entrez) 1027
Concentration 0.2mg/mL
Host Species Mouse
Quantity 0.5mL