Please login to your online account to display your discounted pricing

anti-p27/Kip1, Clone: SX53G8, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone SX53G8
Host Species Mouse
Isotype κ
Immunogen Purified GST-p27 fusion protein of human origin
Antigen p27/Kip1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2328891M

 
novus biologicals canada
NBP2328891ML
Unlabeled 1mL Each for $429.00

NBP232889

 
novus biologicals canada
NBP232889
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone SX53G8
Host Species Mouse
Isotype κ
Immunogen Purified GST-p27 fusion protein of human origin
Antigen p27/Kip1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P46527
Gene ID (Entrez) 1027
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL