Please login to your online account to display your discounted pricing

anti-Osteoprotegerin/TNFRSF11B, Clone: 98A1071, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 98A1071
Host Species Mouse
Isotype IgG1, κ
Immunogen A peptide corresponding to the amino acids 20-37 TQETFPPKYLHYDEETSH of human Osteoprotegerin was used as immunogen for this antibody.
Antigen Osteoprotegerin/TNFRSF11B
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056505T

 
novus biologicals canada
NB10056505SS
Unlabeled 0.025mg Each for $139.00

NB10056505

 
novus biologicals canada
NB10056505
Unlabeled 0.1mg Each for $329.00
Description & Specifications

Specifications

Clone 98A1071
Host Species Mouse
Isotype IgG1, κ
Immunogen A peptide corresponding to the amino acids 20-37 TQETFPPKYLHYDEETSH of human Osteoprotegerin was used as immunogen for this antibody.
Antigen Osteoprotegerin/TNFRSF11B
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity The Osteoprotegerin antibody, clone 98A1071, detects all forms of Osteoprotegerin including monomeric, dimeric and Osteoprotegerin bound to RANKL (Vandooren, 2008) as well as Osteoprotegerin bound to TRAIL (Sandra, 2006).
Gene Accession No. O00300
Gene ID (Entrez) 4982
Formulation PBS containing 0.05% BSA.
Concentration 0.5mg/mL