Please login to your online account to display your discounted pricing

anti-Ornithine Decarboxylase, Clone: ODC1/485, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232887

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232887

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Isotype κ
Immunogen Recombinant human ODC-1 protein
Quantity 0.5mL
Formulation PBS with 0.05% BSA.
Clone ODC1/485
Gene Accession No. P11926
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4953
Concentration 0.2mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Test Specificity Recognizes a 53kDa protein, identified as the Ornithine Decarboxylase (ODC-1). ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC-1 is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater than those in adenomas and normal mucosa.
Antigen Ornithine Decarboxylase
Host Species Mouse
Regulatory Status RUO
Format Purified