Please login to your online account to display your discounted pricing

anti-Ornithine Decarboxylase, Clone: ODC1/485, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone ODC1/485
Host Species Mouse
Isotype κ
Immunogen Recombinant human ODC-1 protein
Antigen Ornithine Decarboxylase
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232887

 
novus biologicals canada
NBP232887
Unlabeled 0.5mL Each for $321.00

NBP2328871M

 
novus biologicals canada
NBP2328871ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone ODC1/485
Host Species Mouse
Isotype κ
Immunogen Recombinant human ODC-1 protein
Antigen Ornithine Decarboxylase
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Recognizes a 53kDa protein, identified as the Ornithine Decarboxylase (ODC-1). ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC-1 is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater than those in adenomas and normal mucosa.
Gene Accession No. P11926
Gene ID (Entrez) 4953
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL