Please login to your online account to display your discounted pricing

anti-Ornithine Decarboxylase, Clone: ODC1/485, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328871ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328871M

  • / Each
 

Description & Specifications

Specifications

Antigen Ornithine Decarboxylase
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone ODC1/485
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P11926
Host Species Mouse
Immunogen Recombinant human ODC-1 protein
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4953
Test Specificity Recognizes a 53kDa protein, identified as the Ornithine Decarboxylase (ODC-1). ODC is the initial and rate-limiting enzyme in the biosynthetic pathway of polyamines and is involved in the conversion of ornithine to putrescine. The biological activity of ODC-1 is rapidly induced in response to virtually all agents known to promote cell proliferation including hormones, drugs, growth factors, mitogens, and tumor promoters. Reportedly, ODC mRNA levels are elevated in lung carcinomas as well as in colon adenomas and carcinomas. ODC activity in colorectal carcinomas is greater than those in adenomas and normal mucosa.