Please login to your online account to display your discounted pricing

anti-OPA1, Clone: 1E8-1D9, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 1E8-1D9
Host Species Murine
Isotype κ
Immunogen Human OPA1 [Swiss-Prot No. O60313].
Antigen OPA1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP171656

 
novus biologicals canada
NBP171656
Unlabeled 0.1mL Each for $329.00

NBP171656SS

 
novus biologicals canada
NBP171656SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone 1E8-1D9
Host Species Murine
Isotype κ
Immunogen Human OPA1 [Swiss-Prot No. O60313].
Antigen OPA1
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Gene Accession No. O60313
Gene ID (Entrez) 4976