Please login to your online account to display your discounted pricing

anti-OCIL/CLEC2d, Clone: 166C1309, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 166C1309
Host Species Mouse
Isotype IgG1
Immunogen GenBank no. gbAAL37198.1.
Antigen OCIL/CLEC2d
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056553

 
novus biologicals canada
NB10056553
Unlabeled 0.1mg Each for $329.00

NB10056553T

 
novus biologicals canada
NB10056553SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 166C1309
Host Species Mouse
Isotype IgG1
Immunogen GenBank no. gbAAL37198.1.
Antigen OCIL/CLEC2d
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene ID (Entrez) 29121
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL