Please login to your online account to display your discounted pricing

anti-NUT, Clone: 2B6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP231147SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP231147SS

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Mouse
Clone 2B6
Quantity 0.025mg
Monoclonal or Polyclonal Monoclonal
Antigen NUT
Immunogen Partial recombinant protein made to an internal portion of human NUT (between amino acids 400-700) [UniProt Q86Y26].
Isotype κ
Regulatory Status RUO
Gene ID (Entrez) 256646
Formulation PBS
Gene Accession No. Q86Y26
Primary or Secondary Primary
Concentration 1.0mg/mL
Conjugate Unlabeled