Please login to your online account to display your discounted pricing

anti-Nucleoli Marker, Clone: NM95, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232886

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232886

  • / Each
 

Description & Specifications

Specifications

Quantity 0.5mL
Antigen Nucleoli Marker
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Immunogen Nuclei of myeloid leukemia biopsy cells
Conjugate Unlabeled
Cross Reactivity Human
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Concentration 0.2mg/mL
Host Species Mouse
Isotype κ
Monoclonal or Polyclonal Monoclonal
Format Purified
Clone NM95