Please login to your online account to display your discounted pricing

anti-Nucleoli Marker, Clone: NM95, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone NM95
Host Species Mouse
Isotype κ
Immunogen Nuclei of myeloid leukemia biopsy cells
Antigen Nucleoli Marker
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232886

 
novus biologicals canada
NBP232886
Unlabeled 0.5mL Each for $321.00

NBP2328861M

 
novus biologicals canada
NBP2328861ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone NM95
Host Species Mouse
Isotype κ
Immunogen Nuclei of myeloid leukemia biopsy cells
Antigen Nucleoli Marker
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL