Please login to your online account to display your discounted pricing

anti-Nucleoli Marker, Clone: NM95, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328861ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328861M

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Immunogen Nuclei of myeloid leukemia biopsy cells
Isotype κ
Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Format Purified
Host Species Mouse
Concentration 0.2mg/mL
Antigen Nucleoli Marker
Quantity 1mL
Clone NM95
Cross Reactivity Human