Please login to your online account to display your discounted pricing

anti-Nuclear Membrane Marker, Clone: NM97, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232885

 View all options for this product


For Research Use Only

Catalog No. NBP232885


Description & Specifications


Antigen Nuclear Membrane Marker
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone NM97
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Nuclei of myeloid leukemia biopsy cells
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal