Please login to your online account to display your discounted pricing

anti-Nuclear Membrane Marker, Clone: NM97, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone NM97
Host Species Mouse
Isotype κ
Immunogen Nuclei of myeloid leukemia biopsy cells
Antigen Nuclear Membrane Marker
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2328851M

 
novus biologicals canada
NBP2328851ML
Unlabeled 1mL Each for $429.00

NBP232885

 
novus biologicals canada
NBP232885
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone NM97
Host Species Mouse
Isotype κ
Immunogen Nuclei of myeloid leukemia biopsy cells
Antigen Nuclear Membrane Marker
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL