Please login to your online account to display your discounted pricing

anti-Nuclear Membrane Marker, Clone: NM97, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328851ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328851M

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Regulatory Status RUO
Quantity 1mL
Antigen Nuclear Membrane Marker
Formulation PBS with 0.05% BSA.
Cross Reactivity Human
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Concentration 0.2mg/mL
Isotype κ
Host Species Mouse
Clone NM97
Immunogen Nuclei of myeloid leukemia biopsy cells