Please login to your online account to display your discounted pricing

anti-Notch-1, Clone: mN1A, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10078486

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10078486

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 0.1mL
Test Specificity Does not cross-react with Notch 2, 3, or 4. Mouse and human. Does not recognize rat Notch1.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4851
Isotype IgG1, κ
Antigen Notch-1
Regulatory Status RUO
Format Purified
Immunogen Mouse Notch1 protein [UniProt Q01705]
Clone mN1A
Host Species Mouse
Concentration 1.0mg/mL
Formulation PBS
Conjugate Unlabeled