Please login to your online account to display your discounted pricing

anti-NLRP3/NALP3, Clone: 8N8E9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203947

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203947

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. NP_001073289.1
Immunogen A recombinant protein corresponding to the Pyrin domain (PYD) of human NALP3 (amino acids 6-89) was used as the immunogen for this antibody. NALP3 belongs to a class of protein motifs known as the death fold. As a class
Isotype κ
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Host Species Mouse
Applications Western Blotting
Regulatory Status RUO
Quantity 0.1mg
Clone 8N8E9
Antigen NLRP3/NALP3
Primary or Secondary Primary
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 114548
Conjugate Unlabeled