Please login to your online account to display your discounted pricing

anti-NLRP3/NALP3, Clone: 8N8E9, Novus Biologicals™

$139.00 - $274.59

Specifications

Clone 8N8E9
Host Species Mouse
Isotype κ
Immunogen A recombinant protein corresponding to the Pyrin domain (PYD) of human NALP3 (amino acids 6-89) was used as the immunogen for this antibody. NALP3 belongs to a class of protein motifs known as the death fold. As a class
Antigen NLRP3/NALP3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP203947

 
novus biologicals canada
NBP203947
Unlabeled 0.1mg Each for $274.59

NBP203947SS

 
novus biologicals canada
NBP203947SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 8N8E9
Host Species Mouse
Isotype κ
Immunogen A recombinant protein corresponding to the Pyrin domain (PYD) of human NALP3 (amino acids 6-89) was used as the immunogen for this antibody. NALP3 belongs to a class of protein motifs known as the death fold. As a class
Antigen NLRP3/NALP3
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene Accession No. NP_001073289.1
Gene ID (Entrez) 114548
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA