Please login to your online account to display your discounted pricing

anti-NLRP3/NALP3, Clone: 8N8E9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP203947SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP203947SS

  • / Each
 

Description & Specifications

Specifications

Antigen NLRP3/NALP3
Applications Western Blot
Clone 8N8E9
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Gene Accession No. NP_001073289.1
Host Species Murine
Immunogen A recombinant protein corresponding to the Pyrin domain (PYD) of human NALP3 (amino acids 6-89) was used as the immunogen for this antibody. NALP3 belongs to a class of protein motifs known as the death fold. As a class
Isotype κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 114548
Target Species Human
Target Species Murine