Please login to your online account to display your discounted pricing

anti-NKX3.1, Clone: 0361, Novus Biologicals™

$144.63 - $384.00

Specifications

Clone 0361
Host Species Murine
Immunogen Full-length human Nkx3.1 produced in bacteria as a hexahistidine-tagged protein. [UniProt No. Q99801]
Antigen NKX3.1
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB1001828SS

 
novus biologicals canada
NB1001828SS
Unlabeled 0.025mL Each for $144.63

NB1001828

 
novus biologicals canada
NB1001828
Unlabeled 0.1mL Each for $384.00
Description & Specifications

Specifications

Clone 0361
Host Species Murine
Immunogen Full-length human Nkx3.1 produced in bacteria as a hexahistidine-tagged protein. [UniProt No. Q99801]
Antigen NKX3.1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Mouse and Rat. Predicted to work in human based on 100% sequence homology.
Gene Accession No. Q99801
Gene ID (Entrez) 4824
Formulation PBS, pH 7.4
Concentration 2.3mg/mL