Please login to your online account to display your discounted pricing

anti-Mouse NKp46/NCR1, PE, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSFAB2225P025

 View more versions of this product

Catalog No. FAB2225P025

  • / Each
 

Description & Specifications

Specifications

Antigen NKp46/NCR1
Format Affinity Purified
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse NKp46/NCR1, Glu22-Asn255, Accession No. Q8C567
Isotype IgG
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 17086
Conjugate PE
Cross Reactivity Mouse
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q8C567