Please login to your online account to display your discounted pricing

anti-Mouse NKp46/NCR1, PE, Polyclonal, R&D Systems

$129.05 - $376.73

Specifications

Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q8C567
Gene ID (Entrez) 17086
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

FAB2225P025

 
r & d systems
RDSFAB2225P025
PE 25 Tests Each for $129.05

FAB2225P100

 
r & d systems
RDSFAB2225P100
PE 100 Tests Each for $376.73
Description & Specifications

Specifications

Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q8C567
Gene ID (Entrez) 17086

Host Species: Goat
Species Reactivity:Mouse
Immunogen:Mouse myeloma cell line NS0-derived recombinant mouse NKp46/NCR1, Glu22-Asn255, Accession No. Q8C567

Applications:

Flow Cytometry