Please login to your online account to display your discounted pricing

anti-Mouse NKp46/NCR1, PE, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSFAB2225P100

 View more versions of this product

Catalog No. FAB2225P100

  • / Each
 

Description & Specifications

Specifications

Antigen NKp46/NCR1
Conjugate PE
Cross Reactivity Mouse
Format Affinity Purified
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q8C567
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse NKp46/NCR1, Glu22-Asn255, Accession No. Q8C567
Isotype IgG
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 17086
Target Species Murine