Please login to your online account to display your discounted pricing

anti-Mouse NKp46/NCR1, Fluorescein, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSFAB2225F100

 View more versions of this product

Catalog No. FAB2225F100

  • / Each
 

Description & Specifications

Specifications

Antigen NKp46/NCR1
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse NKp46/NCR1, Glu22-Asn255, Accession No. Q8C567
Host Species Goat
Quantity 100 Tests
Cross Reactivity Mouse
Format Affinity Purified
Regulatory Status RUO
Isotype IgG
Conjugate Fluorescein
Primary or Secondary Primary
Gene ID (Entrez) 17086
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Accession No. Q8C567
Monoclonal or Polyclonal Polyclonal