Please login to your online account to display your discounted pricing

anti-NHE3/SLC9A3, Clone: 19F5, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11061586

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11061586

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Rabbit
Cross Reactivity Rat
Antigen NHE3/SLC9A3
Quantity 0.1mL
Clone 19F5
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Format Purified
Formulation Tris-glycine, 150mM NaCl
Gene ID (Entrez) 6550
Test Specificity This antibody is specific for NHE3. It does not show any cross-reactivity with any other NHE isoforms. Rat and rabbit. Immunogen sequence has 83% homology to mouse. Does not react with human.
Immunogen Fusion protein representing the C-terminal 131 amino acids of rabbit NHE3. [UniProt No. P26432]
Regulatory Status RUO
Host Species Mouse
Isotype IgG1
Conjugate Unlabeled