Please login to your online account to display your discounted pricing

anti-NHE3/SLC9A3, Clone: 19F5, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 19F5
Host Species Mouse
Isotype IgG1
Immunogen Fusion protein representing the C-terminal 131 amino acids of rabbit NHE3. [UniProt No. P26432]
Antigen NHE3/SLC9A3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11061586

 
novus biologicals canada
NB11061586
Unlabeled 0.1mL Each for $329.00

NB11061586T

 
novus biologicals canada
NB11061586SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone 19F5
Host Species Mouse
Isotype IgG1
Immunogen Fusion protein representing the C-terminal 131 amino acids of rabbit NHE3. [UniProt No. P26432]
Antigen NHE3/SLC9A3
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This antibody is specific for NHE3. It does not show any cross-reactivity with any other NHE isoforms. Rat and rabbit. Immunogen sequence has 83% homology to mouse. Does not react with human.
Gene ID (Entrez) 6550
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL