Please login to your online account to display your discounted pricing

anti-NHE3/SLC9A3, Clone: 19F5, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11061586SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11061586T

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Fusion protein representing the C-terminal 131 amino acids of rabbit NHE3. [UniProt No. P26432]
Antigen NHE3/SLC9A3
Monoclonal or Polyclonal Monoclonal
Test Specificity This antibody is specific for NHE3. It does not show any cross-reactivity with any other NHE isoforms. Rat and rabbit. Immunogen sequence has 83% homology to mouse. Does not react with human.
Concentration 1mg/mL
Cross Reactivity Rabbit
Cross Reactivity Rat
Gene ID (Entrez) 6550
Conjugate Unlabeled
Primary or Secondary Primary
Formulation Tris-glycine, 150mM NaCl
Quantity 0.025mL
Clone 19F5
Isotype IgG1
Host Species Mouse
Regulatory Status RUO