Please login to your online account to display your discounted pricing

anti-NFkB p105/p50, Clone: 2J10D7, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056583

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056583

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Quantity 0.1mg
Isotype IgG1, κ
Gene ID (Entrez) 4790
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Gene Accession No. P25942
Conjugate Unlabeled
Host Species Mouse
Test Specificity Cross reacts with Human. Predicted to react with Mouse and Rat which are 86% homologous with the immunogen.
Cross Reactivity Human
Formulation PBS containing 0.05% BSA
Antigen NFkB p105/p50
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A portion of amino acids 150-200 of human NF-kB (p50) was used as the immunogen.
Regulatory Status RUO
Clone 2J10D7
Format Purified