Please login to your online account to display your discounted pricing

anti-Nestin, Clone: 10C2, Novus Biologicals™


Manufacturer: novus biologicals canada  NB300266SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB300266SS

  • / Each
 

Description & Specifications

Specifications

Antigen Nestin
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone 10C2
Concentration 2.4mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS
Gene Accession No. P48681
Host Species Murine
Immunogen A fragment of the human Nestin protein corresponding to residues 1464-1614. [UniProt P48681]
Isotype IgG1, κ
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 10763
Test Specificity Human. It does not cross-react mouse, rat or rabbit.
Target Species Human