Please login to your online account to display your discounted pricing

anti-NAC1, Clone: 3, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11077345SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11077345T

  • / Each
 

Description & Specifications

Specifications

Antigen NAC1
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone 3
Concentration 0.82mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. Q96RE7
Host Species Murine
Immunogen Recombinant human Nac1 protein. [UniProt No. Q96RE7]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 112939
Test Specificity Human and rat. Mouse reactivity reported in scientific literature (PMID: 23922682).
Target Species Human
Target Species Murine
Target Species Rat