Please login to your online account to display your discounted pricing

anti-NAC1, Clone: 3, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11077345

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11077345

  • / Each
 

Description & Specifications

Specifications

Immunogen Recombinant human Nac1 protein. [UniProt No. Q96RE7]
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen NAC1
Test Specificity Human and rat. Mouse reactivity reported in scientific literature (PMID: 23922682).
Gene ID (Entrez) 112939
Gene Accession No. Q96RE7
Host Species Mouse
Clone 3
Concentration 0.82mg/mL
Monoclonal or Polyclonal Monoclonal
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Regulatory Status RUO
Primary or Secondary Primary
Formulation Tris-glycine, 150mM NaCl
Conjugate Unlabeled