Please login to your online account to display your discounted pricing

anti-NAC1, Clone: 3, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 3
Host Species Murine
Immunogen Recombinant human Nac1 protein. [UniProt No. Q96RE7]
Antigen NAC1
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11077345T

 
novus biologicals canada
NB11077345SS
Unlabeled 0.025mL Each for $109.00

NB11077345

 
novus biologicals canada
NB11077345
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Clone 3
Host Species Murine
Immunogen Recombinant human Nac1 protein. [UniProt No. Q96RE7]
Antigen NAC1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity Human and rat. Mouse reactivity reported in scientific literature (PMID: 23922682).
Gene Accession No. Q96RE7
Gene ID (Entrez) 112939
Formulation Tris-glycine, 150mM NaCl
Concentration 0.82mg/mL