Please login to your online account to display your discounted pricing

anti-n-Myc, Clone: NMYC-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NB200109

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB200109

  • / Each
 

Description & Specifications

Specifications

Applications Chromatin Immunoprecipitation
Applications Gel Supershift
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Immunogen A synthetic peptide (SPYVESEDAPPQKC) conjugated to KLH
Test Specificity This does not react with c-Myc or L-Myc. Human and mouse. Not yet tested in other species.
Gene ID (Entrez) 4613
Clone NMYC-1
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. P04198
Concentration 1.0mg/mL
Primary or Secondary Primary
Regulatory Status RUO
Isotype IgG2a, κ
Format Purified
Quantity 0.1mL
Antigen n-Myc
Host Species Mouse
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal