Please login to your online account to display your discounted pricing

anti-N-Cadherin, Clone: 13A9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP148309

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP148309

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Antigen N-Cadherin
Regulatory Status RUO
Formulation Tris-glycine, 150mM NaCl
Conjugate Unlabeled
Format Purified
Gene ID (Entrez) 1000
Concentration 1mg/mL
Primary or Secondary Primary
Gene Accession No. P19022
Isotype IgG1
Host Species Mouse
Clone 13A9
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen Cytoplasmic domain of human N Cadherin [Swiss-Prot No. P19022]
Monoclonal or Polyclonal Monoclonal