Please login to your online account to display your discounted pricing

anti-Myogenin, Clone: F5D, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232986

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232986

  • / Each
 

Description & Specifications

Specifications

Antigen Myogenin
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone F5D
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P15173
Host Species Mouse
Immunogen Human myogenin recombinant protein
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4656