Please login to your online account to display your discounted pricing

anti-Myogenin, Clone: F5D, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone F5D
Host Species Mouse
Isotype κ
Immunogen Human myogenin recombinant protein
Antigen Myogenin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232986

 
novus biologicals canada
NBP232986
Unlabeled 0.5mL Each for $321.00

NBP2329861M

 
novus biologicals canada
NBP2329861ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone F5D
Host Species Mouse
Isotype κ
Immunogen Human myogenin recombinant protein
Antigen Myogenin
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P15173
Gene ID (Entrez) 4656
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL