Please login to your online account to display your discounted pricing

anti-Myogenin, Clone: F5D, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329861ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329861M

  • / Each
 

Description & Specifications

Specifications

Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 1mL
Antigen Myogenin
Immunogen Human myogenin recombinant protein
Regulatory Status RUO
Format Purified
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Gene Accession No. P15173
Gene ID (Entrez) 4656
Isotype κ
Primary or Secondary Primary
Clone F5D
Host Species Mouse