Please login to your online account to display your discounted pricing

anti-MyoD, Clone: MYD712, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2328831ML

 View all options for this product


For Research Use Only

Catalog No. NBP2328831M


Description & Specifications


Antigen MyoD
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone MYD712
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P15172
Host Species Mouse
Immunogen Recombinant human MyoD1 protein
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4654