Please login to your online account to display your discounted pricing

anti-MyoD, Clone: MYD712, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232883

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232883

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Gene Accession No. P15172
Primary or Secondary Primary
Gene ID (Entrez) 4654
Conjugate Unlabeled
Format Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype κ
Clone MYD712
Antigen MyoD
Regulatory Status RUO
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Quantity 0.5mL
Host Species Mouse
Immunogen Recombinant human MyoD1 protein