Please login to your online account to display your discounted pricing

anti-MyoD, Clone: MYD712, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone MYD712
Host Species Mouse
Isotype κ
Immunogen Recombinant human MyoD1 protein
Antigen MyoD
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2328831M

 
novus biologicals canada
NBP2328831ML
Unlabeled 1mL Each for $429.00

NBP232883

 
novus biologicals canada
NBP232883
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone MYD712
Host Species Mouse
Isotype κ
Immunogen Recombinant human MyoD1 protein
Antigen MyoD
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P15172
Gene ID (Entrez) 4654
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL