Please login to your online account to display your discounted pricing

anti-MyoD, Clone: 5.8A, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056511SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056511T

  • / Each
 

Description & Specifications

Specifications

Antigen MyoD
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blot
Clone 5.8A
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA
Gene Accession No. P15172
Host Species Murine
Immunogen The 5.8A antibody was made against recombinant mouse MyoD protein but it also recognizes human (myf3)
Isotype IgG1, κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4654
Test Specificity In Rh-30, a ∽45 kDa band should be observed. Cat, Human, Mouse, Rat. Not yet tested in other species.
Target Species Feline
Target Species Human
Target Species Murine
Target Species Rat