Please login to your online account to display your discounted pricing

anti-MyD88, Clone: 4D6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227369

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227369

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 4615
Test Specificity The amino acid sequence used as immunogen is 100% homologous in Human (isoform 1: 317 amino acids , isoform 3: 264 aa, isoform 4: 204 aa; isoform 5: 159 aa, CRA_a: 304 aa, CRA_b: 309 aa), Gorilla, Chimp, Rhesus Monkey and Orangutan, 82% homologous in Horse, 73% homologous in Dog, 68% homologous in Cat and Mouse (isoform CRA_a: 314 amino acids, CRA_b: 177 amino acids), and 63% homologous in Rat.
Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal
Gene Accession No. Q99836
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Quantity 0.1mg
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Format Purified
Clone 4D6
Immunogen A portion of amino acids 50-100 of human MyD88 was used as the immunogen for this antibody.
Regulatory Status RUO
Host Species Mouse
Antigen MyD88
Conjugate Unlabeled