Please login to your online account to display your discounted pricing

anti-MyD88, Clone: 4D6, Novus Biologicals™

$139.00 - $384.00

Specifications

Clone 4D6
Host Species Murine
Immunogen A portion of amino acids 50-100 of human MyD88 was used as the immunogen for this antibody.
Antigen MyD88
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227369

 
novus biologicals canada
NBP227369
Unlabeled 0.1mg Each for $384.00

NBP227369SS

 
novus biologicals canada
NBP227369SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 4D6
Host Species Murine
Immunogen A portion of amino acids 50-100 of human MyD88 was used as the immunogen for this antibody.
Antigen MyD88
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in Human (isoform 1: 317 amino acids , isoform 3: 264 aa, isoform 4: 204 aa; isoform 5: 159 aa, CRA_a: 304 aa, CRA_b: 309 aa), Gorilla, Chimp, Rhesus Monkey and Orangutan, 82% homologous in Horse, 73% homologous in Dog, 68% homologous in Cat and Mouse (isoform CRA_a: 314 amino acids, CRA_b: 177 amino acids), and 63% homologous in Rat.
Gene Accession No. Q99836
Gene ID (Entrez) 4615
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL