Please login to your online account to display your discounted pricing

anti-MyD88, Clone: 4D6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227369SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227369SS

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Immunogen A portion of amino acids 50-100 of human MyD88 was used as the immunogen for this antibody.
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Clone 4D6
Host Species Mouse
Format Purified
Quantity 0.025mg
Regulatory Status RUO
Antigen MyD88
Primary or Secondary Primary
Conjugate Unlabeled
Gene ID (Entrez) 4615
Gene Accession No. Q99836
Concentration 0.5mg/mL
Test Specificity The amino acid sequence used as immunogen is 100% homologous in Human (isoform 1: 317 amino acids , isoform 3: 264 aa, isoform 4: 204 aa; isoform 5: 159 aa, CRA_a: 304 aa, CRA_b: 309 aa), Gorilla, Chimp, Rhesus Monkey and Orangutan, 82% homologous in Horse, 73% homologous in Dog, 68% homologous in Cat and Mouse (isoform CRA_a: 314 amino acids, CRA_b: 177 amino acids), and 63% homologous in Rat.
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Monoclonal or Polyclonal Monoclonal