Please login to your online account to display your discounted pricing

anti-Muscleblind-like 1, Clone: HL-1822 3A4-1E9, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11037256

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11037256

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Host Species Mouse
Gene ID (Entrez) 4154
Conjugate Unlabeled
Primary or Secondary Primary
Gene Accession No. Q9NR56
Immunogen Recombinant human Muscleblind-like 1 protein. [UniProt No. Q9NR56]
Clone HL-1822 3A4-1E9
Antigen Muscleblind-like 1
Isotype IgG1, κ
Quantity 0.5mL
Format Supernatant
Monoclonal or Polyclonal Monoclonal