Please login to your online account to display your discounted pricing

anti-Muscleblind-like 1, Clone: HL-1822 3A4-1E9, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone HL-1822 3A4-1E9
Host Species Murine
Isotype IgG1, κ
Immunogen Recombinant human Muscleblind-like 1 protein. [UniProt No. Q9NR56]
Antigen Muscleblind-like 1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11037256

 
novus biologicals canada
NB11037256
Unlabeled 0.5mL Each for $329.00

NB11037256T

 
novus biologicals canada
NB11037256SS
Unlabeled 0.1mL Each for $139.00
Description & Specifications

Specifications

Clone HL-1822 3A4-1E9
Host Species Murine
Isotype IgG1, κ
Immunogen Recombinant human Muscleblind-like 1 protein. [UniProt No. Q9NR56]
Antigen Muscleblind-like 1
Monoclonal or Polyclonal Monoclonal
Format Supernatant
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. Q9NR56
Gene ID (Entrez) 4154